Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNDBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012142.4 | NP_036274.3 | 7 | 794 | ||
NR_027513.2 | 5 | 617 | ||||
NR_027514.2 | 6 | 697 | ||||
NR_045998.1 | 6 | 714 | ||||
NR_045999.1 | 4 | 557 | ||||
XM_006720448.1 | XP_006720511.1 | 5 | 716 | |||
GenBank mRNA | AF082569.1 | - | 7 | 653 | ||
AF087852.1 | - | 7 | 639 | |||
AF113535.1 | - | 5 | 603 | |||
AF132034.1 | - | 7 | 609 | |||
AF246144.1 | - | 7 | 611 | |||
AK075146.1 | - | 7 | 623 | |||
AK075296.1 | - | 5 | 608 | |||
AK124193.1 | - | 2 | 1198 | |||
AK128849.1 | - | 3 | 1838 | |||
AK225143.1 | - | 7 | 609 | |||
AK225645.1 | - | 7 | 623 | |||
AK290665.1 | - | 7 | 623 | |||
AK290707.1 | - | 6 | 576 | |||
AK295017.1 | - | 6 | 526 | |||
AK308817.1 | - | 6 | 1218 | |||
AL552334.3 | - | 7 | 733 | |||
BC009689.1 | - | 7 | 637 | 3896421 | ||
BI911845.1 | - | 468 | ||||
BQ439439.1 | - | 4 | 371 | |||
CR450331.1 | - | 7 | 563 |