Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193534.1 | NP_001180463.1 | 2 | 415 | ||
NM_001193535.1 | NP_001180464.1 | 2 | 364 | |||
NM_012161.3 | NP_036293.1 | 2 | 415 | |||
NR_036464.1 | 2 | 415 | ||||
XM_006713959.3 | XP_006714022.1 | 3 | 511 | |||
XM_011513831.2 | XP_011512133.1 | 2 | 197 | |||
XM_011513832.2 | XP_011512134.1 | 2 | 344 | |||
XM_017008019.1 | XP_016863508.1 | 4 | 1282 | |||
GenBank mRNA | AF142481.1 | - | 2 | 337 | ||
AF157323.1 | - | 2 | 276 | |||
AF176700.1 | - | 2 | 299 | |||
AF199420.1 | - | 2 | 304 | |||
AK000153.1 | - | 1 | 125 | |||
AK295510.1 | - | 2 | 334 | |||
AK310860.1 | - | 2 | 344 | |||
AL832808.1 | - | 1 | 147 | |||
BC012320.1 | - | 2 | 344 | |||
BC030656.1 | - | 2 | 386 | 4837080 | ||
BI464829.1 | - | 2 | 396 | |||
BX537957.1 | - | 2 | 310 |