Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faslg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012908.1 | NP_037040.1 | 4 | 882 | ||
XM_006250148.2 | XP_006250210.1 | 7 | 1521 | |||
XM_008769607.1 | XP_008767829.1 | 8 | 1756 | |||
GenBank mRNA | U03470.1 | - | 4 | 882 |