Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012923.2 | NP_037055.1 | 6 | 1197 | ||
GenBank mRNA | BC081852.1 | - | 6 | 1168 | 7105829 | |
FQ211577.1 | - | 5 | 1170 | |||
FQ211975.1 | - | 6 | 1249 | |||
FQ224502.1 | - | 6 | 1158 | |||
X70871.1 | - | 6 | 1167 |