Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012932.2 | NP_037064.1 | 14 | 2015 | ||
XM_006251084.2 | XP_006251146.1 | 14 | 2249 | |||
GenBank mRNA | U52102.1 | - | 14 | 1983 |