Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278732.1 | NP_001265661.1 | 2, 3 | 514 | ||
NM_012208.3 | NP_036340.1 | 2, 3 | 414 | |||
XM_011537619.1 | XP_011535921.1 | 3, 4 | 427 | |||
XM_011537620.1 | XP_011535922.1 | 3, 4 | 427 | |||
XM_017009287.1 | XP_016864776.1 | 2, 3 | 414 | |||
XM_017009288.1 | XP_016864777.1 | 3, 4 | 478 | |||
XM_017009289.1 | XP_016864778.1 | 3, 4 | 471 | |||
XM_017009290.1 | XP_016864779.1 | 2, 3 | 464 | |||
XM_017009291.1 | XP_016864780.1 | 2, 3 | 157 | |||
XM_017009292.1 | XP_016864781.1 | 2, 3 | 101 | |||
GenBank mRNA | AK293269.1 | - | 2, 3 | 294 | ||
AK309832.1 | - | 2, 3 | 512 | |||
AK311277.1 | - | 2, 3 | 510 | |||
AK312976.1 | - | 2, 3 | 232 | |||
BC007680.2 | - | 2, 3 | 233 | 3628006 | ||
BC014982.1 | - | 2, 3 | 378 | 4859849 | ||
BQ228600.1 | - | 2, 3 | 248 | |||
DA150729.1 | - | 2, 3 | 416 | |||
U18937.1 | - | 2, 3 | 340 |