Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAT4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012214.2 | NP_036346.1 | 4 | 680 | ||
GenBank mRNA | AB000616.1 | - | 3 | 501 | ||
AK291533.1 | - | 3 | 490 | |||
AK314024.1 | - | 4 | 644 | |||
BC127107.1 | - | 3 | 578 | 40124731 | ||
BC127108.1 | - | 3 | 578 | 40124741 |