Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013048.2 | NP_037180.1 | 2 | 277 | ||
XM_008763588.1 | XP_008761810.1 | 4 | 284 | |||
XM_008763589.1 | XP_008761811.1 | 3 | 227 |