Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013113.2 | NP_037245.2 | 3 | 441 | ||
GenBank mRNA | BC078902.1 | - | 3 | 441 | 7111949 | |
J02701.1 | - | 3 | 764 | |||
M14137.1 | - | 3 | 345 |