Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnlip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013161.2 | NP_037293.1 | 9 | 884 | ||
GenBank mRNA | D88534.1 | - | 8 | 856 | ||
D88535.1 | - | 8 | 856 | |||
M58369.1 | - | 9 | 885 |