Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPSA SIRNA Assays ›
Gene Symbol:
Rpsa
Gene Name:
ribosomal protein SA
Gene Aliases:
Lamr1
Chromosome Location:
Chr.8: 128806056 - 128809987 on Build Rnor_6.0
UniGene ID:
Rn.121972
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
198121
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rpsa
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017138.2
NP_058834.1
3
243
XM_008766672.1
XP_008764894.1
3
316
GenBank mRNA
BC060578.1
-
3
261
6919235
D25224.1
-
3
244
FJ386454.1
-
2
224
FQ209463.1
-
3
257
FQ209469.1
-
3
257
FQ209490.1
-
3
258
FQ209667.1
-
3
257
FQ209729.1
-
3
257
FQ211189.1
-
3
257
FQ212660.1
-
3
257
FQ212893.1
-
3
257
FQ213275.1
-
3
257
FQ213317.1
-
3
257
FQ213334.1
-
3
257
FQ213623.1
-
3
260
FQ213835.1
-
3
258
FQ213894.1
-
3
257
FQ215242.1
-
3
257
FQ215292.1
-
3
258
FQ216063.1
-
3
257
FQ216085.1
-
3
257
FQ216130.1
-
3
257
FQ216245.1
-
3
257
FQ216788.1
-
3
259
FQ216997.1
-
3
257
FQ217664.1
-
3
257
FQ219586.1
-
3
257
FQ219785.1
-
3
260
FQ220012.1
-
3
257
FQ220017.1
-
3
255
FQ220075.1
-
3
258
FQ220084.1
-
3
260
FQ220126.1
-
3
257
FQ220134.1
-
3
257
FQ220171.1
-
3
256
FQ220184.1
-
3
257
FQ220347.1
-
3
259
FQ220461.1
-
3
238
FQ220468.1
-
3
257
FQ220661.1
-
3
260
FQ220822.1
-
3
257
FQ220904.1
-
3
258
FQ220964.1
-
3
257
FQ221103.1
-
3
257
FQ221334.1
-
3
277
FQ221443.1
-
3
258
FQ221672.1
-
3
257
FQ222093.1
-
3
259
FQ222280.1
-
3
257
FQ222542.1
-
3
259
FQ223223.1
-
3
257
FQ224747.1
-
3
257
FQ225510.1
-
3
258
FQ226054.1
-
3
257
FQ226408.1
-
3
257
FQ227056.1
-
3
257
FQ227293.1
-
3
257
FQ227600.1
-
3
258
FQ228002.1
-
3
257
FQ228212.1
-
3
258
FQ229290.1
-
3
259
FQ229305.1
-
3
259
FQ229367.1
-
3
258
FQ229516.1
-
3
257
FQ229579.1
-
3
257
FQ229598.1
-
3
258
FQ229712.1
-
3
257
FQ229787.1
-
3
257
FQ229850.1
-
3
259
FQ229859.1
-
3
255
FQ229890.1
-
3
257
FQ229918.1
-
3
277
FQ229970.1
-
3
257
FQ230088.1
-
3
257
FQ230180.1
-
3
258
FQ230258.1
-
3
256
FQ231469.1
-
2
74
FQ231622.1
-
3
255
FQ232305.1
-
3
259
FQ232568.1
-
3
258
FQ233068.1
-
3
257
FQ233112.1
-
3
257
FQ233340.1
-
3
259
FQ233547.1
-
3
258
FQ233919.1
-
3
257
FQ234511.1
-
3
257
FQ234742.1
-
3
257
FQ234765.1
-
3
257
FQ234772.1
-
3
257
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal small subunit assembly
endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
rRNA export from nucleus
translation
single organismal cell-cell adhesion
epithelial cell differentiation
structural constituent of ribosome
laminin receptor activity
structural molecule activity
ribosome binding
laminin binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0