Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012203.1 | NP_036335.1 | 9 | 1149 | ||
XM_005251631.1 | XP_005251688.1 | 6 | 842 | |||
XM_011518073.2 | XP_011516375.1 | 9 | 1523 | |||
GenBank mRNA | AF113215.1 | - | 9 | 1157 | ||
AF113251.1 | - | 9 | 1167 | |||
AF134895.1 | - | 9 | 1149 | |||
AF146018.1 | - | 9 | 1148 | |||
AK024386.1 | - | 2 | 4675 | |||
AK025978.1 | - | 9 | 1193 | |||
AK026287.1 | - | 4 | 1506 | |||
AK026752.1 | - | 4 | 1810 | |||
BC000605.2 | - | 9 | 1128 | 3347232 | ||
BC003131.1 | - | 9 | 1149 | |||
BC088360.1 | - | 9 | 1147 |