Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahcy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017201.1 | NP_058897.1 | 5, 6 | 592 | ||
GenBank mRNA | BC061841.1 | - | 5, 6 | 619 | 5599994 | |
M15185.1 | - | 5, 6 | 592 |