Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017253.2 | NP_058949.1 | 2 | 205 | ||
XM_008763370.1 | XP_008761592.1 | 2 | 218 | |||
XM_008763371.1 | XP_008761593.1 | 2 | 468 | |||
XM_008763372.1 | XP_008761594.1 | 3 | 294 | |||
XM_008763373.1 | XP_008761595.1 | 2 | 397 | |||
GenBank mRNA | AJ278701.1 | - | 2 | 199 | ||
BC087710.1 | - | 2 | 205 | 7315507 | ||
U35774.1 | - | 2 | 171 |