Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000369.2 | NP_000360.2 | 4 | 499 | ||
NM_001018036.2 | NP_001018046.1 | 4 | 499 | |||
NM_001142626.2 | NP_001136098.1 | 4 | 499 | |||
XM_005268037.4 | XP_005268094.1 | 5 | 719 | |||
XM_005268039.1 | XP_005268096.1 | 5 | 719 | |||
XM_006720245.1 | XP_006720308.1 | 5 | 719 | |||
XM_011537119.2 | XP_011535421.1 | 4 | 444 | |||
GenBank mRNA | AB209207.1 | - | 4 | 371 | ||
AY429111.1 | - | 4 | 343 | |||
BC009237.2 | - | 4 | 408 | |||
BC024205.2 | - | 4 | 593 | |||
BC063613.1 | - | 4 | 375 | |||
BC108653.1 | - | 4 | 416 | |||
BC120972.2 | - | 4 | 369 | |||
BC120973.2 | - | 4 | 369 | |||
BC127628.1 | - | 4 | 369 | |||
BC141970.1 | - | 4 | 369 | |||
M31774.1 | - | 4 | 443 | |||
M32215.1 | - | 4 | 387 | |||
M73747.1 | - | 4 | 413 | |||
S45272.1 | - | 4 | 343 | |||
S49816.1 | - | 4 | 364 |