Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321196.1 | NP_001308125.1 | 10 | 1774 | ||
NM_012254.2 | NP_036386.1 | 10 | 2026 | |||
XM_011526364.1 | XP_011524666.1 | 9 | 1824 | |||
GenBank mRNA | AF064255.1 | - | 10 | 1986 | ||
AK026640.1 | - | 3 | 292 | |||
AK123036.1 | - | 10 | 2026 | |||
AK298446.1 | - | 10 | 1701 | |||
BX103174.1 | - | 3 | 168 |