Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017289.1 | NP_058985.1 | 6 | 729 | ||
GenBank mRNA | BC087714.1 | - | 6 | 694 | 7317154 | |
M35162.1 | - | 6 | 729 |