Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019122.1 | NP_061995.1 | 3 | 588 | ||
XM_006228978.2 | XP_006229040.1 | 4 | 1046 | |||
XM_006228979.2 | XP_006229041.1 | 4 | 1051 | |||
XM_008759324.1 | XP_008757546.1 | 5 | 776 | |||
XM_008759325.1 | XP_008757547.1 | 4 | 680 | |||
GenBank mRNA | AF375464.1 | - | 2 | 486 | ||
BC088145.1 | - | 5 | 801 | |||
D28512.1 | - | 3 | 588 | |||
EU441216.1 | - | 2 | 491 |