Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019158.2 | NP_062031.1 | 6 | 980 | ||
XM_006230187.2 | XP_006230249.1 | 6 | 1027 | |||
GenBank mRNA | AB005547.1 | - | 6 | 979 | ||
AF007775.1 | - | 6 | 959 | |||
BC081812.1 | - | 6 | 980 | 7114305 |