Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019211.2 | NP_062084.2 | 2 | 286 | ||
GenBank mRNA | AF060819.1 | - | 2 | 148 | ||
AF081196.1 | - | 2 | 286 |