Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coro1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019222.1 | NP_062095.1 | 5 | 618 | ||
XM_006230773.2 | XP_006230835.1 | 5 | 737 | |||
XM_006230774.2 | XP_006230836.1 | 5 | 702 | |||
GenBank mRNA | AJ006064.1 | - | 5 | 618 | ||
BC061558.1 | - | 5 | 671 | 6919414 |