Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021587.1 | NP_067598.1 | 2, 3 | 883 | ||
GenBank mRNA | M55431.1 | - | 2, 3 | 883 |