Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012401.3 | NP_036533.2 | 25 | 4297 | ||
XM_005261909.1 | XP_005261966.1 | 26 | 4345 | |||
XM_005261910.2 | XP_005261967.1 | 26 | 4356 | |||
XM_005261911.4 | XP_005261968.1 | 25 | 4295 | |||
XM_006724413.2 | XP_006724476.1 | 27 | 4415 | |||
XM_011530682.2 | XP_011528984.1 | 26 | 4496 | |||
XM_017028703.1 | XP_016884192.1 | 27 | 4396 | |||
XM_017028704.1 | XP_016884193.1 | 26 | 4349 | |||
GenBank mRNA | AB002313.1 | - | 24 | 4214 | ||
AB075511.1 | - | 10 | 1407 | |||
AK025701.1 | - | 12 | 1665 | |||
AK056543.1 | - | 8 | 1466 | |||
AK074932.1 | - | 7 | 933 | |||
AK126394.1 | - | 6 | 1006 | |||
BC004542.2 | - | 2 | 116 | |||
BC148225.1 | - | 3 | 476 | |||
BC148280.1 | - | 3 | 476 | |||
BT006887.1 | - | 2 | 100 | |||
S76730.1 | - | 3 | 449 |