Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031585.1 | NP_113773.1 | 3, 4 | 339 | ||
XM_008769222.1 | XP_008767444.1 | 3, 4 | 447 | |||
GenBank mRNA | D00036.1 | - | 3, 4 | 339 |