Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012294.3 | NP_036426.3 | 12, 13 | 1131 | ||
XM_005249914.2 | XP_005249971.1 | 12, 13 | 1350 | |||
XM_005249916.1 | XP_005249973.1 | 12, 13 | 1294 | |||
XM_011515652.2 | XP_011513954.1 | 4, 5 | 3104 | |||
XM_017012837.1 | XP_016868326.1 | 10, 11 | 1035 | |||
GenBank mRNA | AK295859.1 | - | 2, 3 | 616 | ||
BC039203.1 | - | 2, 3 | 442 | 4811483 | ||
BP349215.1 | - | 3, 4 | 189 | |||
D87467.1 | - | 2, 3 | 423 | |||
KU178490.1 | - | 2, 3 | 368 | |||
KU178491.1 | - | 2, 3 | 368 |