Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080478.1 | NP_536726.1 | 3, 4 | 965 | ||
XM_006229935.1 | XP_006229997.1 | 4, 5 | 1334 | |||
XM_006229936.2 | XP_006229998.1 | 4, 5 | 1170 | |||
XM_006229938.2 | XP_006230000.1 | 4, 5 | 1509 | |||
XM_006229940.2 | XP_006230002.1 | 3, 4 | 350 | |||
XM_006229941.2 | XP_006230003.1 | 3, 4 | 384 | |||
XM_008759717.1 | XP_008757939.1 | 3, 4 | 349 | |||
GenBank mRNA | AF333983.1 | - | 3, 4 | 965 | ||
X60469.1 | - | 3, 4 | 660 |