Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Llgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246518.2 | XP_006246580.1 | 18, 19 | 2926 | ||
XM_006246519.2 | XP_006246581.1 | 18, 19 | 2923 | |||
GenBank mRNA | AF356187.1 | - | 18, 19 | 2771 | ||
FQ228753.1 | - | 1, 2 | 77 |