Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172036.3 | NP_742033.1 | 3 | 255 | ||
GenBank mRNA | AF161588.1 | - | 3 | 269 | ||
BC058441.1 | - | 3 | 259 | 6918795 | ||
FQ224284.1 | - | 3 | 221 | |||
FQ229256.1 | - | 3 | 257 |