Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012522.2 | NP_036654.2 | 10 | 1120 | ||
GenBank mRNA | D01098.1 | - | 10 | 1121 | ||
FQ211441.1 | - | 10 | 1179 | |||
M88344.1 | - | 10 | 1269 | |||
M88345.1 | - | 7 | 1533 | |||
M88346.1 | - | 9 | 1015 | |||
M88347.1 | - | 9 | 1015 |