Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012339.3 | NP_036471.1 | 4 | 524 | ||
XM_005269666.4 | XP_005269723.1 | 2 | 984 | |||
XM_005269667.3 | XP_005269724.1 | 2 | 267 | |||
XM_011539562.2 | XP_011537864.1 | 2 | 227 | |||
XM_011539563.2 | XP_011537865.1 | 2 | 438 | |||
XM_017016010.1 | XP_016871499.1 | 4 | 530 | |||
GenBank mRNA | AF120266.1 | - | 4 | 515 | ||
BC003157.1 | - | 4 | 518 | 2958221 | ||
BC004161.2 | - | 4 | 518 | 2958221 |