Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012605.2 | NP_036737.1 | 4 | 269 | ||
GenBank mRNA | CB770854.1 | - | 4 | 279 | ||
FQ215085.1 | - | 4 | 302 | |||
FQ216891.1 | - | 4 | 285 | |||
FQ216934.1 | - | 4 | 285 | |||
FQ217090.1 | - | 4 | 287 | |||
FQ217098.1 | - | 4 | 285 | |||
FQ217115.1 | - | 2 | 134 | |||
FQ217155.1 | - | 4 | 285 | |||
FQ217219.1 | - | 4 | 285 | |||
FQ217259.1 | - | 4 | 285 | |||
FQ217349.1 | - | 3 | 209 | |||
FQ217365.1 | - | 4 | 284 | |||
FQ217368.1 | - | 4 | 301 | |||
FQ217703.1 | - | 4 | 285 | |||
FQ217740.1 | - | 4 | 301 | |||
FQ217881.1 | - | 4 | 282 | |||
FQ217944.1 | - | 4 | 285 | |||
FQ218017.1 | - | 4 | 285 | |||
FQ223762.1 | - | 4 | 266 | |||
FQ223868.1 | - | 4 | 285 | |||
FQ223911.1 | - | 4 | 301 | |||
FQ223944.1 | - | 3 | 151 | |||
FQ223946.1 | - | 4 | 285 | |||
FQ223966.1 | - | 4 | 285 | |||
FQ223985.1 | - | 4 | 285 | |||
FQ224150.1 | - | 2 | 128 | |||
FQ224231.1 | - | 4 | 285 | |||
FQ224253.1 | - | 4 | 284 | |||
FQ224391.1 | - | 4 | 283 | |||
FQ224404.1 | - | 4 | 283 | |||
FQ224511.1 | - | 4 | 285 | |||
FQ224525.1 | - | 4 | 304 | |||
FQ224720.1 | - | 4 | 285 | |||
FQ224781.1 | - | 4 | 301 | |||
J00754.1 | - | 1 | 21 |