Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239906.2 | 4, 5 | 645 | |||
XM_006239907.2 | XP_006239969.1 | 4, 5 | 635 | |||
XM_006239908.2 | XP_006239970.1 | 3, 4 | 551 | |||
XM_008764573.1 | 4, 5 | 618 | ||||
GenBank mRNA | BC078882.1 | - | 2, 3 | 323 | 7115603 | |
J04791.1 | - | 4, 5 | 439 | |||
M16982.1 | - | 4, 5 | 703 |