Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034111.1 | NP_001029283.1 | 5 | 401 | ||
NM_012630.2 | NP_036762.1 | 5 | 411 | |||
XM_008760751.1 | XP_008758973.1 | 5 | 1000 | |||
GenBank mRNA | BC092133.1 | - | 5 | 405 | 7310645 | |
L48060.1 | - | 305 | ||||
M19304.1 | - | 5 | 412 | |||
M34083.1 | - | 4 | 297 | |||
M57668.1 | - | 5 | 401 | |||
M74152.1 | - | 5 | 371 | |||
U07567.1 | - | 5 | 354 |