Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012663.2 | NP_036795.1 | 3 | 339 | ||
XM_006246593.2 | XP_006246655.1 | 3 | 757 | |||
GenBank mRNA | AJ133104.1 | - | 3 | 323 | ||
BC074003.1 | - | 3 | 339 | 7097683 | ||
DQ521403.1 | - | 1 | 80 | |||
M24105.1 | - | 2 | 277 |