Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016998.3 | NP_058694.2 | 3 | 545 | ||
GenBank mRNA | FQ232505.1 | - | 3 | 253 | ||
J00713.1 | - | 3 | 545 | |||
V01232.1 | - | 3 | 248 |