Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012294.3 | NP_036426.3 | 26 | 3494 | ||
XM_005249914.2 | XP_005249971.1 | 26 | 3713 | |||
XM_005249916.1 | XP_005249973.1 | 26 | 3657 | |||
XM_011515652.2 | XP_011513954.1 | 18 | 5467 | |||
XM_017012837.1 | XP_016868326.1 | 24 | 3398 | |||
GenBank mRNA | BC039203.1 | - | 14 | 2398 | 4811483 | |
D87467.1 | - | 16 | 2786 |