Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAGI2 SIRNA Assays ›
Gene Symbol:
MAGI2
Gene Name:
membrane associated guanylate kinase, WW and PDZ domain containing 2
Gene Aliases:
ACVRIP1, AIP-1, AIP1, ARIP1, MAGI-2, SSCAM
Chromosome Location:
Chr.7: 78017055 - 79453805 on Build GRCh38
UniGene ID:
Hs.603842
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
20059
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAGI2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301128.1
NP_001288057.1
17, 18
3410
NM_012301.3
NP_036433.2
18, 19
3452
XM_011516718.1
XP_011515020.1
18, 19
3683
XM_011516719.2
XP_011515021.1
18, 19
7100
XM_011516720.2
XP_011515022.1
18, 19
12090
XM_011516726.2
XP_011515028.1
15, 16
2756
XM_011516728.1
XP_011515030.1
14, 15
2292
XM_017012840.1
XP_016868329.1
19, 20
3812
XM_017012841.1
XP_016868330.1
19, 20
3809
XM_017012842.1
XP_016868331.1
19, 20
3806
XM_017012843.1
XP_016868332.1
18, 19
3770
XM_017012844.1
XP_016868333.1
19, 20
3812
XM_017012845.1
XP_016868334.1
18, 19
3677
XM_017012846.1
XP_016868335.1
17, 18
3641
XM_017012847.1
XP_016868336.1
19, 20
10049
XM_017012848.1
XP_016868337.1
17, 18
6968
XM_017012849.1
XP_016868338.1
17, 18
6962
XM_017012850.1
XP_016868339.1
19, 20
3812
XM_017012851.1
XP_016868340.1
19, 20
3812
XM_017012852.1
XP_016868341.1
19, 20
3812
GenBank mRNA
AB014605.2
-
17, 18
3391
AF038563.1
-
18, 19
3452
AK297353.1
-
18, 19
3437
AK309012.1
-
13, 14
2246
BC150277.1
-
17, 18
3391
9007255
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001170746, NM_001170745, NM_015823, AK122349, AK147530, and others.
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of receptor internalization
planar cell polarity pathway involved in axis elongation
negative regulation of cell proliferation
positive regulation of neuron projection development
negative regulation of cell migration
positive regulation of phosphoprotein phosphatase activity
negative regulation of activin receptor signaling pathway
nerve growth factor signaling pathway
receptor clustering
protein heterooligomerization
negative regulation of protein kinase B signaling
SMAD protein signal transduction
mitotic cell cycle arrest
glomerular visceral epithelial cell development
neuroligin clustering involved in postsynaptic membrane assembly
cellular response to nerve growth factor stimulus
positive regulation of synaptic vesicle clustering
signal transducer activity
protein binding
phosphatase binding
receptor signaling complex scaffold activity
beta-1 adrenergic receptor binding
SMAD binding
type II activin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0