Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017048.2 | NP_058744.1 | 4 | 441 | ||
XM_006235874.2 | XP_006235936.1 | 4 | 898 | |||
XM_006235875.2 | XP_006235937.1 | 4 | 619 | |||
XM_006235876.2 | XP_006235938.1 | 4 | 681 | |||
XM_006235877.2 | XP_006235939.1 | 3 | 824 | |||
XM_006235878.2 | XP_006235940.1 | 3 | 743 | |||
GenBank mRNA | AJ288902.1 | - | 3 | 236 | ||
J05166.1 | - | 4 | 445 |