Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cth | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017074.1 | NP_058770.1 | 2, 3 | 261 | ||
GenBank mRNA | D17370.1 | - | 2, 3 | 261 | ||
X53460.1 | - | 149 |