Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256468.1 | NP_001243397.1 | 5, 6 | 1544 | ||
NM_001256469.1 | NP_001243398.1 | 5, 6 | 1544 | |||
NM_012330.3 | NP_036462.2 | 5, 6 | 1544 | |||
XM_005269664.2 | XP_005269721.1 | 5, 6 | 1605 | |||
XM_017016000.1 | XP_016871489.1 | 5, 6 | 1453 | |||
XM_017016001.1 | XP_016871490.1 | 5, 6 | 1317 | |||
XM_017016002.1 | XP_016871491.1 | 5, 6 | 1556 | |||
XM_017016003.1 | XP_016871492.1 | 4, 5 | 1227 | |||
XM_017016004.1 | XP_016871493.1 | 5, 6 | 1453 | |||
XM_017016005.1 | XP_016871494.1 | 5, 6 | 1453 | |||
XM_017016006.1 | XP_016871495.1 | 5, 6 | 1453 | |||
XM_017016007.1 | XP_016871496.1 | 5, 6 | 1316 | |||
XM_017016008.1 | XP_016871497.1 | 4, 5 | 1227 | |||
XM_017016009.1 | XP_016871498.1 | 5, 6 | 1544 | |||
GenBank mRNA | AB002381.2 | - | 3, 4 | 843 | ||
AF113514.1 | - | 4, 5 | 1158 | |||
AF119230.1 | - | 4, 5 | 1158 | |||
AF119231.1 | - | 4, 5 | 1158 | |||
AF217500.1 | - | 5, 6 | 1331 | |||
AK074816.1 | - | 5, 6 | 1357 | |||
BC014143.2 | - | 5, 6 | 1336 | |||
BC048199.1 | - | 5, 6 | 1336 | |||
BC150270.1 | - | 3, 4 | 870 | 9007238 | ||
BC150618.1 | - | 5, 6 | 1304 | 9056988 |