Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271077.1 | NP_001258006.1 | 5, 6 | 647 | ||
NM_001271078.1 | NP_001258007.1 | 5, 6 | 284 | |||
NM_019154.1 | NP_062027.1 | 5, 6 | 638 | |||
GenBank mRNA | U01245.2 | - | 4, 5 | 606 | ||
U07054.1 | - | 4, 5 | 225 | |||
U67130.1 | - | 5, 6 | 638 |