Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF508795.1 | - | 3 | 297 | ||
BC062403.1 | - | 3 | 269 | 5621394 | ||
CK480694.1 | - | 3 | 262 | |||
FQ209590.1 | - | 3 | 280 | |||
FQ218571.1 | - | 3 | 282 | |||
FQ220674.1 | - | 3 | 280 | |||
M15327.1 | - | 3 | 253 | |||
X72792.1 | - | 1 | 83 |