Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021838.2 | NP_068610.1 | 11 | 1486 | ||
XM_006235872.1 | XP_006235934.1 | 11 | 1515 | |||
GenBank mRNA | AB176831.1 | - | 11 | 1486 | ||
AJ011115.1 | - | 1 | 64 | |||
AY695391.1 | - | 12 | 1420 |