Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PVALB SIRNA Assays ›
Gene Symbol:
Pvalb
Gene Name:
parvalbumin
Gene Aliases:
PALB1, Pva
Chromosome Location:
Chr.7: 119428652 - 119443656 on Build Rnor_6.0
UniGene ID:
Rn.2005
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
200951
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pvalb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022499.2
NP_071944.1
2
98
XM_006241929.2
XP_006241991.1
3
171
GenBank mRNA
BC126090.1
-
2
98
7321065
FQ212120.1
-
3
161
FQ214597.1
-
3
159
FQ214667.1
-
3
160
FQ214909.1
-
3
159
FQ215072.1
-
3
159
FQ215172.1
-
3
159
FQ215250.1
-
3
160
FQ215353.1
-
3
159
FQ215367.1
-
3
159
FQ215388.1
-
3
159
FQ215455.1
-
3
159
FQ215487.1
-
3
159
FQ215608.1
-
3
160
FQ215681.1
-
3
159
FQ216147.1
-
3
159
FQ216186.1
-
3
159
FQ216201.1
-
3
160
FQ216338.1
-
3
160
FQ216339.1
-
3
159
FQ216448.1
-
3
159
FQ216459.1
-
3
159
FQ216659.1
-
3
159
FQ216789.1
-
3
159
FQ216830.1
-
3
159
FQ216843.1
-
3
159
FQ216867.1
-
3
159
FQ216872.1
-
3
159
FQ216903.1
-
3
161
FQ216906.1
-
3
156
FQ216960.1
-
3
159
FQ216981.1
-
3
159
FQ217053.1
-
3
159
FQ217064.1
-
3
159
FQ217099.1
-
3
159
FQ217132.1
-
2
160
FQ217209.1
-
3
158
FQ217210.1
-
3
159
FQ217278.1
-
3
159
FQ217298.1
-
3
160
FQ217358.1
-
3
159
FQ217373.1
-
3
160
FQ217384.1
-
3
159
FQ217418.1
-
3
160
FQ217424.1
-
3
159
FQ217446.1
-
2
127
FQ217489.1
-
3
159
FQ217527.1
-
3
159
FQ217562.1
-
3
159
FQ217576.1
-
3
159
FQ217645.1
-
3
158
FQ217652.1
-
3
160
FQ217677.1
-
3
160
FQ217743.1
-
3
158
FQ217749.1
-
3
160
FQ217760.1
-
3
160
FQ217770.1
-
3
159
FQ217787.1
-
3
160
FQ217845.1
-
3
159
FQ217858.1
-
3
159
FQ217860.1
-
3
160
FQ217861.1
-
3
159
FQ217880.1
-
3
161
FQ217886.1
-
3
159
FQ217934.1
-
2
101
FQ217966.1
-
3
159
FQ217976.1
-
3
159
FQ217991.1
-
3
159
FQ218022.1
-
3
159
FQ218035.1
-
3
159
FQ218057.1
-
3
159
FQ223582.1
-
3
159
FQ223596.1
-
3
159
FQ223603.1
-
3
160
FQ223630.1
-
3
156
FQ223664.1
-
3
160
FQ223695.1
-
3
159
FQ223712.1
-
3
160
FQ223798.1
-
3
161
FQ223861.1
-
3
159
FQ223863.1
-
3
159
FQ223909.1
-
3
159
FQ223941.1
-
3
159
FQ223961.1
-
3
159
FQ223988.1
-
3
158
FQ223989.1
-
3
159
FQ223990.1
-
3
159
FQ224014.1
-
3
159
FQ224025.1
-
3
160
FQ224028.1
-
3
159
FQ224037.1
-
3
159
FQ224053.1
-
3
159
FQ224059.1
-
3
159
FQ224083.1
-
3
159
FQ224113.1
-
3
159
FQ224173.1
-
3
159
FQ224181.1
-
3
159
FQ224242.1
-
3
160
FQ224248.1
-
3
157
FQ224294.1
-
3
159
FQ224303.1
-
3
160
FQ224322.1
-
3
159
FQ224356.1
-
3
159
FQ224363.1
-
3
159
FQ224417.1
-
3
159
FQ224430.1
-
3
159
FQ224435.1
-
3
157
FQ224463.1
-
3
160
FQ224483.1
-
3
159
FQ224485.1
-
3
159
FQ224488.1
-
3
160
FQ224510.1
-
3
159
FQ224541.1
-
3
159
FQ224580.1
-
3
159
FQ224584.1
-
3
160
FQ224600.1
-
3
159
FQ224613.1
-
3
159
FQ224634.1
-
3
159
FQ224643.1
-
3
159
FQ224673.1
-
3
160
FQ224675.1
-
3
159
FQ224723.1
-
3
161
FQ224767.1
-
3
159
FQ224773.1
-
3
161
FQ224778.1
-
3
159
FQ224913.1
-
3
159
M12725.1
-
3
158
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013645, BC027424, X54613, S75909, S75910, and others.
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytosolic calcium ion concentration
calcium ion binding
protein homodimerization activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0