Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024001.1 | NP_076491.1 | 2 | 284 | ||
XM_006253937.2 | XP_006253999.1 | 2 | 402 | |||
GenBank mRNA | AF288289.1 | - | 2 | 284 | ||
BC083554.1 | - | 2 | 260 | |||
BC098916.1 | - | 2 | 259 |