Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030995.1 | NP_112257.1 | 4 | 3279 | ||
XM_006234826.2 | XP_006234888.1 | 5 | 3643 | |||
XM_006234827.2 | XP_006234889.1 | 5 | 3643 | |||
XM_006234828.2 | XP_006234890.1 | 4 | 3547 | |||
XM_008762145.1 | XP_008760367.1 | 4 | 3283 | |||
GenBank mRNA | M83196.1 | - | 4 | 3279 | ||
X66840.1 | - | 1 | 2609 |