Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012413.3 | NP_036545.1 | 6 | 1042 | ||
GenBank mRNA | AK290605.1 | - | 6 | 1024 | ||
BC036721.1 | - | 6 | 921 | 5224222 | ||
BC047756.2 | - | 6 | 1019 | 6058335 | ||
X67731.1 | - | 5 | 724 | |||
X71125.1 | - | 6 | 931 |