Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031572.1 | NP_113760.1 | 4, 5 | 634 | ||
GenBank mRNA | BC089790.1 | - | 4, 5 | 642 | 7371667 | |
FQ209627.1 | - | 1, 2 | 73 | |||
FQ219038.1 | - | 2, 3 | 315 | |||
FQ219491.1 | - | 3, 4 | 509 | |||
FQ219720.1 | - | 1, 2 | 104 | |||
J03786.1 | - | 4, 5 | 634 |