Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012437.5 | NP_036569.1 | 4 | 760 | ||
NR_052019.1 | 3 | 713 | ||||
NR_052020.1 | 3 | 699 | ||||
GenBank mRNA | AK024555.1 | - | 4 | 737 | ||
BC000761.2 | - | 4 | 689 | 2821705 | ||
BC004494.2 | - | 4 | 690 | 3830132 | ||
BE278772.1 | - | 3 | 615 |