Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp21a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057101.2 | NP_476442.2 | 10 | 1953 | ||
XM_006255907.2 | XP_006255969.1 | 10 | 2027 | |||
GenBank mRNA | U56853.1 | - | 10 | 1954 | ||
U56854.1 | - | 7 | 2169 |